Novus Biologicals
Manufacturer Code:NBP300245DL594
Catalog # NB065045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PRTFDC1 (Q9NRG1-1 Met1-Val225). |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.2.8 FLJ11888 HHGPEC 2.4.2 phosphoribosyl transferase domain containing 1 phosphoribosyltransferase domain-containing protein 1; Phosphoribosyltransferase domain-containing protein 1
Gene Aliases: HHGP; PRTFDC1
UniProt ID: (Human) Q9NRG1
Entrez Gene ID: (Human) 56952
Molecular Function:
glycosyltransferase
isomerase
mutase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.