Proteintech
Manufacturer Code:155271AP150UL
Catalog # 50-173-0289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRSS8 Fusion Protein Ag7234 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP1; CAP1 PROSTASIN protease serine 8 PRSS8 Serine protease 8; channel activating protease 1; Channel-activating protease 1; Prostasin; Prostasin heavy chain; Prostasin light chain; protease, serine, 8 (prostasin); Serine protease 8
Gene Aliases: 2410039E18Rik; AI313909; C79772; CAP1; fr; mCAP1; PROSTASIN; PRSS8
UniProt ID: (Human) Q16651, (Rat) Q9ES87
Entrez Gene ID: (Human) 5652, (Mouse) 76560, (Rat) 192107
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.