Invitrogen
Manufacturer Code:PA548034
Catalog # PIPA548034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Trypsin 3 Phe16-Asn246 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: mesotrypsin; MTG PRSS4 T9 TRY3 TRY4 brain trypsinogen mesotrypsin mesotrypsinogen pancreatic trypsinogen III protease serine 4 (trypsin 4 brain) trypsin III trypsin IV trypsin-3 trypsinogen 4 trypsinogen 5 trypsinogen IV; protease, serine, 3; trypsin 3; trypsinogen 15
Gene Aliases: MTG; Tb; Try3; TRY4
Entrez Gene ID: (Mouse) 22073
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.