Invitrogen
Manufacturer Code:PA5114077
Catalog # PIPA5114077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human PRSS3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain trypsinogen; Brain trypsinogen Mesotrypsin Mesotrypsinogen MTG pancreatic trypsinogen III protease serine 3 protease serine 3 protease serine 4 (trypsin 4 brain) PRSS3 PRSS4 Serine protease 3 Serine protease 4 T9 Tb TRY3 TRY4 trypsin 3 Trypsin III Trypsin IV Trypsin-3 trypsinogen trypsinogen 15 trypsinogen 4 trypsinogen 5 trypsinogen IV; Mesotrypsin; Mesotrypsinogen; pancreatic trypsinogen III; protease, serine, 4 (trypsin 4, brain); Serine protease 3; Serine protease 4; Trypsin III; Trypsin IV; Trypsin-3; trypsinogen 4; trypsinogen 5; trypsinogen IV
Gene Aliases: MTG; PRSS3; PRSS4; T9; TRY3; TRY4
UniProt ID: (Human) P35030
Entrez Gene ID: (Human) 5646
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.