OriGene
Manufacturer Code:CF809867
Catalog # 50-166-9759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9H8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 152-268 of human PRRT2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BFIC2 BFIS2 DSPB3 DYT10 EKD1 FICCA ICCA IFITMD1 PKC; Dispanin subfamily B member 3; DSPB3; dystonia 10; infantile convulsions and paroxysmal choreoathetosis; interferon induced transmembrane protein domain containing 1; Proline-rich transmembrane protein 2
Gene Aliases: BFIC2; BFIS2; DSPB3; DYT10; EKD1; FICCA; ICCA; IFITMD1; PKC; PRRT2
UniProt ID: (Human) Q7Z6L0
Entrez Gene ID: (Human) 112476
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.