Invitrogen
Manufacturer Code:PA534537
Catalog # PIPA534537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the center of human PRRT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dispanin subfamily B member 3; DSPB3; dystonia 10; infantile convulsions and paroxysmal choreoathetosis; interferon induced transmembrane protein domain containing 1; Proline-rich transmembrane protein 2; PRRT2 Proline-rich transmembrane protein 2 dispanin subfamily B member 3 DSPB3 interferon induced transmembrane protein domain containing 1 IFITMD1 DYT10 EKD1
Gene Aliases: 1500031I19Rik; AI195361; BFIC2; BFIS2; DSPB3; DYT10; EKD1; FICCA; ICCA; IFITMD1; PKC; PRRT2; RGD1564195
UniProt ID: (Human) Q7Z6L0, (Mouse) E9PUL5, (Rat) D3ZFB6
Entrez Gene ID: (Human) 112476, (Mouse) 69017, (Rat) 361651
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.