Invitrogen
Manufacturer Code:PA569650
Catalog # PIPA569650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human PRRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dispanin subfamily D member 1; DSPD1; interferon induced transmembrane protein domain containing 7; Proline-rich transmembrane protein 1; Proline-rich transmembrane protein 1 Dispanin subfamily D member 1 DSPD1 PRRT1 C6orf31 NG5; prrt1 {ECO:0000250|UniProtKB:Q99946}; Synapse differentiation-induced protein 4; SynDIG4
Gene Aliases: Bat-4; Bat4; C6orf31; C7H6orf31; D17H6S54E; DSPD1; G5b; IFITMD7; NG5; ORF31; PRRT1
UniProt ID: (Human) Q99946, (Rat) Q6MG82, (Mouse) O35449
Entrez Gene ID: (Rabbit) 100342890, (Dog) 607660, (Bovine) 617717, (Human) 80863, (Horse) 100051413, (Pig) 100144520, (Rat) 406167, (Mouse) 260297
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.