Invitrogen
Manufacturer Code:PA5111480
Catalog # PIPA5111480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human PRRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bat4 Bat-4 BOS_21650 C6orf31 C7H6orf31 D17H6S54E Dispanin subfamily D member 1 DSPD1 G5b IFITMD7 NG5 Orf31 Proline-rich transmembrane protein 1 PRRT1 SBAB-649D6.1 Synapse differentiation-induced protein 4 SynDIG4; Dispanin subfamily D member 1; DSPD1; interferon induced transmembrane protein domain containing 7; Proline-rich transmembrane protein 1; Synapse differentiation-induced protein 4; SynDIG4
Gene Aliases: C6orf31; DSPD1; IFITMD7; NG5; PRRT1
UniProt ID: (Human) Q99946
Entrez Gene ID: (Human) 80863
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.