Novus Biologicals
Manufacturer Code:NBP230641
Catalog # NBP230641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PAQTAQAPGFVVPTHAGTVGTLPLGGYVAPGYPLQLQPCTAYVPVYPVGTPYAGGTPGGTGVTSTL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf31 Chromosome 6 Open Reading Frame 31 Dispanin Subfamily D Member 1 DSPD1 IFITMD7 Interferon Induced Transmembrane Protein Domain Containing 7 NG5 Proline-Rich Transmembrane Protein 1; Dispanin subfamily D member 1; DSPD1; interferon induced transmembrane protein domain containing 7; Proline-rich transmembrane protein 1; Synapse differentiation-induced protein 4; SynDIG4
Gene Aliases: C6orf31; DSPD1; IFITMD7; NG5; PRRT1
UniProt ID: (Human) Q99946
Entrez Gene ID: (Human) 80863
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.