Invitrogen
Manufacturer Code:PA532185
Catalog # PIPA532185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 58 and 242 of Human Protor1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20185k PP610 PROTOR-1 PROTOR1 proline-rich protein 5 protein observed with Rictor-1; proline rich 5 (renal); Proline-rich protein 5; Protein observed with Rictor-1; Protor-1; prr5; Rho GTPase activating protein 8
Gene Aliases: Arhgap8; AU043908; C030017C09Rik; C78947; FLJ20185k; PP610; PROTOR-1; PROTOR1; PRR5
UniProt ID: (Human) P85299, (Mouse) Q812A5
Entrez Gene ID: (Human) 55615, (Mouse) 109270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.