Invitrogen
Manufacturer Code:PA588677
Catalog # PIPA588677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-240 of human PRPS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Phophoribosylpyrophosphate synthetase, subunit II; Phosphoribosyl pyrophosphate synthase II; phosphoribosyl pyrophosphate synthetase II; PPRibP; PPRibP synthetase; PRS-II; ribose-phosphate diphosphokinase 2; Ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41
Gene Aliases: 2610101M19Rik; AA589463; AI464149; Prps-2; PRPS2; PRSII
UniProt ID: (Human) P11908, (Rat) P09330, (Mouse) Q9CS42
Entrez Gene ID: (Human) 5634, (Rat) 24689, (Mouse) 110639
Molecular Function: carbohydrate kinase kinase ligase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.