Novus Biologicals
Manufacturer Code:NBP15766920UL
Catalog # NBP15766920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PRPS2(phosphoribosyl pyrophosphate synthetase 2) The peptide sequence was selected from the middle region of PRPS2. Peptide sequence VSPDAGGAKRVTSIADRLNVEFALIHKERKKANEVDRMVLVGDVKDRVAI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.6.1 Phosphoribosyl pyrophosphate synthase II phosphoribosyl pyrophosphate synthetase 2 PPRibP PPRibP synthetase PRSII PRS-II ribose-phosphate diphosphokinase 2 ribose-phosphate pyrophosphokinase 2; Phosphoribosyl pyrophosphate synthase II; PPRibP; PPRibP synthetase; PRS-II; ribose-phosphate diphosphokinase 2; Ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41
Gene Aliases: PRPS2; PRSII
UniProt ID: (Human) P11908
Entrez Gene ID: (Human) 5634
Molecular Function: carbohydrate kinase kinase ligase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.