Novus Biologicals
Manufacturer Code:NBP242649G
Catalog # NBP242649G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 100000000000 |
Immunogen | Recombinant human PRPS1 (1-318aa) purified from E. coli |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMTX5ARTS deafness X-linked 2 perceptive congenital DFN2 DFNX1KIAA0967 dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1) EC 2.7.6.1 Phosphoribosyl pyrophosphate synthase I phosphoribosyl pyrophosphate synthetase 1 PPRibP PRSI PRS-I ribose-phosphate diphosphokinase 1 ribose-phosphate pyrophosphokinase 1; deafness 2, perceptive, congenital; deafness, X-linked 2, perceptive, congenital; dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1); Phosphoribosyl pyrophosphate synthase I; PPRibP; PRS-I; ribose-phosphate diphosphokinase 1; Ribose-phosphate pyrophosphokinase 1
Gene Aliases: ARTS; CMTX5; DFN2; DFNX1; PPRibP; PRPS1; PRS-I; PRSI
UniProt ID: (Human) P60891
Entrez Gene ID: (Human) 5631
Molecular Function: carbohydrate kinase kinase ligase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.