Novus Biologicals
Manufacturer Code:NBP222273
Catalog # NBP222273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1825 and 1875 of human Pre-mRNA-Processing Factor 8 using the numbering given in entry NP_006436.3 (GeneID 10594). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 1 apoptosis-regulated protein 2 p220 precursor mRNA processing protein pre-mRNA-processing-splicing factor 8 Prp8 PRP8 homolog PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae) PRP8 pre-mRNA processing factor 8 homolog (yeast) PRP8220 kDa U5 snRNP-specific protein PRPC8HPRP8 RP13 Splicing factor Prp8 U5 snRNP-specific protein (220 kD) ortholog of S. cerevisiae Prp8p; apoptosis-regulated protein 2; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; Splicing factor Prp8; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: HPRP8; PRP8; PRPC8; PRPF8; RP13; SNRNP220
UniProt ID: (Human) Q6P2Q9
Entrez Gene ID: (Human) 10594
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.