Invitrogen
Manufacturer Code:PA518611
Catalog # PIPA518611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KELGNSLDKCKNNEN) corresponding to the internal amino acids of PRPF31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hPrp31; Pre-mRNA-processing factor 31; Protein 61K; PRP31 pre-mRNA processing factor 31 homolog; PRPF31 PRP31 pre-mRNA processing factor 31 homolog (S. cerevisiae) DKFZp566J153 NY-BR-99 PRP31 RP11 PRP31 pre-mRNA processing factor 31 homolog (yeast) pre-mRNA processing factor 31 homolog pre-mRNA processing factor 31 homolog (yeast); Serologically defined breast cancer antigen NY-BR-99; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein
Gene Aliases: NY-BR-99; PRP31; PRPF31; RP11; SNRNP61
UniProt ID: (Human) Q8WWY3
Entrez Gene ID: (Human) 26121
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.