Abnova
Manufacturer Code:H00026121K
Catalog # 89-202-110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PRPF31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PRPF31 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp566J153 NY-BR-99 PRP31 RP11; hPrp31; Pre-mRNA-processing factor 31; Protein 61K; PRP31 pre-mRNA processing factor 31 homolog; Serologically defined breast cancer antigen NY-BR-99; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein
Gene Aliases: NY-BR-99; PRP31; PRPF31; RP11; SNRNP61
UniProt ID: (Human) Q8WWY3
Entrez Gene ID: (Human) 26121
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.