Invitrogen
Manufacturer Code:PA537022
Catalog # PIPA537022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 223-268 of human PRPF18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPRP18; HPRP18 homolog; Potassium channel regulatory factor; Pre-mRNA-splicing factor 18; Pre-mRNA-splicing factor 18 PRP18 homolog hPRP18 PRP18 HPRP18; PRP18 homolog; PRP18 pre-mRNA processing factor 18 homolog; PRP18 pre-mRNA processing factor 18 homolog(PRPF18)
Gene Aliases: 2810441A10Rik; HPRP18; Kcrf; Pprf18; PRP18; PRPF18
UniProt ID: (Human) Q99633, (Rat) Q9JKB8, (Mouse) Q8BM39
Entrez Gene ID: (Human) 8559, (Rat) 171552, (Mouse) 67229
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.