Invitrogen
Manufacturer Code:MA534736
Catalog # PIMA534736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG57-38 |
Immunogen | Recombinant protein within 1-200 of human PRPF8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; 220 kDa U5 snRNP-specific protein AU019467 BOS_18541 D11Bwg0410e DBF3/PRP8 H PRPC8 HPRP8 id:ibd1257 ik:tdsubc_2a9 im:7141966 p220 Pre-mRNA-processing-splicing factor 8 Prp8 PRP8 homolog PRPC8 Prpf8 RP13 Sfprp8l SNRNP220 spl Splicing factor Prp8 tdsubc_2a9 wu:fb37c02 wu:fb73e06 xx:tdsubc_2a9 zgc:56504; apoptosis-regulated protein 1; apoptosis-regulated protein 2; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; Splicing factor Prp8; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: HPRP8; PRP8; PRPC8; PRPF8; RP13; SNRNP220
UniProt ID: (Human) Q6P2Q9
Entrez Gene ID: (Human) 10594
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.