Invitrogen
Manufacturer Code:PA576193
Catalog # PIPA576193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of human PRP8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; PRP8 RP13 HPRP8 PRPC8 SNRNP220; Splicing factor Prp8; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: AU019467; D11Bwg0410e; DBF3/PRP8; HPRP8; PRP8; PRPC8; PRPF8; RP13; Sfprp8l; SNRNP220
UniProt ID: (Human) Q6P2Q9, (Mouse) Q99PV0
Entrez Gene ID: (Human) 10594, (Rat) 287530, (Mouse) 192159
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.