Invitrogen
Manufacturer Code:PA541817
Catalog # PIPA541817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PRPF8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; PRP8 pre-mRNA processing factor 8 homolog (S. cerevisiae) HPRP8 PRP8 PRPC8 RP13; Splicing factor Prp8; U5 snRNP-specific protein; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: AU019467; D11Bwg0410e; DBF3/PRP8; HPRP8; id:ibd1257; ik:tdsubc_2a9; im:7141966; PRP8; PRPC8; PRPF8; RP13; Sfprp8l; SNRNP220; tdsubc_2a9; wu:fb37c02; wu:fb73e06; xx:tdsubc_2a9; zgc:56504
UniProt ID: (Human) Q6P2Q9, (Mouse) Q99PV0
Entrez Gene ID: (Human) 10594, (Rabbit) 100357344, (Bovine) 507371, (Dog) 480651, (Horse) 100072388, (Rat) 287530, (Guinea pig) 100712742, (Mouse) 192159, (Zebrafish) 393951
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.