Invitrogen
Manufacturer Code:PA528721
Catalog # PIPA528721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1978 and 2072 of Human PRP8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; HPRP8 PRP8 PRPC8 RP13 220 kDa U5 snRNP-specific protein PRP8 homolog U5 snRNP-specific protein (220 kD) ortholog of S. cerevisiae Prp8p apoptosis-regulated protein 1 apoptosis-regulated protein 2 p220 pre-mRNA-processing-splicing factor 8 precursor mRNA processing protein spl; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; Splicing factor Prp8; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: HPRP8; PRP8; PRPC8; PRPF8; RP13; SNRNP220
UniProt ID: (Human) Q6P2Q9
Entrez Gene ID: (Human) 10594
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.