Invitrogen
Manufacturer Code:PA5106369
Catalog # PIPA5106369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRPF8 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 220 kDa U5 snRNP-specific protein; 220 kDa U5 snRNP-specific protein apoptosis-regulated protein 1 apoptosis-regulated protein 2 AU019467 D11Bwg0410e DBF3/PRP8 H PRPC8 HPRP8 p220 precursor mRNA processing protein pre-mRNA processing factor 8 pre-mRNA-processing-splicing factor 8 Prp8 PRP8 homolog PRP8 pre-mRNA processing factor 8 homolog PRPC8 Prpf8 RP13 Sfprp8l SNRNP220 spl Splicing factor Prp8 U5 snRNP-specific protein (220 kD) ortholog of S. cerevisiae Prp8p; apoptosis-regulated protein 1; apoptosis-regulated protein 2; p220; Pre-mRNA-processing-splicing factor 8; precursor mRNA processing protein; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; Splicing factor Prp8; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p
Gene Aliases: AU019467; D11Bwg0410e; DBF3/PRP8; HPRP8; PRP8; PRPC8; PRPF8; RP13; Sfprp8l; SNRNP220
UniProt ID: (Human) Q6P2Q9, (Mouse) Q99PV0
Entrez Gene ID: (Human) 10594, (Rat) 287530, (Mouse) 192159
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.