Invitrogen
Manufacturer Code:PA586910
Catalog # PIPA586910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 165-209 of Human PRPF19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPso4; Neuronal differentiation-related gene protein; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; Pre-mRNA-processing factor 19; Pre-mRNA-processing factor 19 Nuclear matrix protein 200 PRP19/PSO4 homolog hPso4 Senescence evasion factor; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor
Gene Aliases: AA617263; AL024362; D19Wsu55e; hPSO4; NMP200; PRP19; PRPF19; PSO4; SNEV; UBOX4
UniProt ID: (Human) Q9UMS4, (Mouse) Q99KP6, (Rat) Q9JMJ4
Entrez Gene ID: (Human) 27339, (Mouse) 28000, (Rat) 246216
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.