Novus Biologicals
Manufacturer Code:NBP192297
Catalog # NBP192297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SNPVLELELAEEKLPMTLSRQEVIRRLRERGEPIRLFGETDYDAFQRLRKIEILTPEVNKGLRNDLKAALDKIDQQYLNEIVG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10210 hPrp18 pre-mRNA-splicing factor 18 PRP18 PRP18 homolog PRP18 pre-mRNA processing factor 18 homolog (S. cerevisiae) PRP18 pre-mRNA processing factor 18 homolog (yeast) PRP18 pre-mRNA processing factor 18 homolog(PRPF18); hPRP18; Pre-mRNA-splicing factor 18; PRP18 homolog; PRP18 pre-mRNA processing factor 18 homolog; PRP18 pre-mRNA processing factor 18 homolog(PRPF18)
Gene Aliases: HPRP18; PRP18; PRPF18
UniProt ID: (Human) Q99633
Entrez Gene ID: (Human) 8559
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.