Invitrogen
Manufacturer Code:PA579886
Catalog # PIPA579886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human PROX1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox prospero-like protein PROX1; Prospero homeobox protein 1; Prospero homeobox protein 1 Homeobox prospero-like protein PROX1 PROX-1 PROX1; prospero-related homeobox 1; PROX-1
Gene Aliases: A230003G05Rik; PROX1
UniProt ID: (Human) Q92786, (Mouse) P48437
Entrez Gene ID: (Human) 5629, (Rat) 305066, (Mouse) 19130
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.