Invitrogen
Manufacturer Code:PA5100890
Catalog # PIPA5100890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PROP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ames dwarf; Ames dwarf CPHD2 df homeobox protein prophet of Pit-1 paired like homeodomain factor 1 pituitary specific homeodomain factor pituitary-specific homeodomain factor PROP paired-like homeobox 1 PROP1 PROP-1 prophet of Pit-1 prophet of Pit1 paired-like homeodomain transcription factor prophet of Pit1 paired-like homeodomain transcription factor; Homeobox protein prophet of Pit-1; paired like homeodomain factor 1; pituitary specific homeodomain factor; Pituitary-specific homeodomain factor; PROP-1; prophet of Pit-1; prophet of Pit1 paired-like homeodomain transcription factor; prophet of Pit1, paired-like homeodomain transcription factor
Gene Aliases: CPHD2; df; PROP-1; PROP1
UniProt ID: (Human) O75360, (Mouse) P97458
Entrez Gene ID: (Human) 5626, (Mouse) 19127, (Rat) 266738
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.