Novus Biologicals
Manufacturer Code:NBP144054
Catalog # NBP144054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence AERRRQAEKPKKGR-C corresponding to N-Terminus according to NP_006252.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CPHD2 homeobox protein prophet of Pit-1 Pituitary-specific homeodomain factor PROP paired-like homeobox 1 PROP-1 prophet of Pit1 paired-like homeodomain transcription factor; Homeobox protein prophet of Pit-1; Pituitary-specific homeodomain factor; PROP-1; prophet of Pit1, paired-like homeodomain transcription factor
Gene Aliases: CPHD2; PROP-1; PROP1
UniProt ID: (Human) O75360
Entrez Gene ID: (Human) 5626
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.