OriGene
Manufacturer Code:TA500315
Catalog # 50-167-1462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G8 |
Immunogen | Full-length protein expressed in 293T cell transfected with human PROM2 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPROML2; PROM-2; Prominin-2; Prominin-like protein 2; prominin-related protein; PROML2
Gene Aliases: PROM2; PROML2; UNQ2521/PRO6014
UniProt ID: (Human) Q8N271
Entrez Gene ID: (Human) 150696
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.