Invitrogen
Manufacturer Code:PA598946
Catalog # PIPA598946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse Prokineticin-2 protein (27-128AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bombina variegata 8 kD protein; Bombina variegata 8kDa protein; Bv 8 homolog (Bombina variegata); PK2; prokineticin 1; Prokineticin-2; Protein Bv8 homolog
Gene Aliases: Bv8; PK2; Prok1; Prok2
UniProt ID: (Mouse) Q9QXU7
Entrez Gene ID: (Mouse) 50501
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.