Invitrogen
Manufacturer Code:PA542039
Catalog # PIPA542039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PRODH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsPOX1; Hydroxyproline dehydrogenase; HYPDH; Kidney and liver proline oxidase 1; MmPOX1; PO; Probable proline dehydrogenase 2; Probable proline oxidase 2; proline dehydrogenase (oxidase) 2; Proline dehydrogenase (oxidase) 2 HSPOX1; proline dehydrogenase 2; proline oxidase 1; Proline oxidase-like protein
Gene Aliases: 2510028N04Rik; 2510038B11Rik; HSPOX1; HYPDH; MmPOX; MmPOX1; POX1; PRODH2
UniProt ID: (Bovine) A6QQ74, (Human) Q9UF12, (Mouse) Q8VCZ9
Entrez Gene ID: (Bovine) 508542, (Pig) 106510519, (Human) 58510, (Dog) 484572, (Mouse) 56189, (Rat) 361538
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.