OriGene
Manufacturer Code:CF810559
Catalog # 50-167-0165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H6 |
Immunogen | Full length human recombit protein of human PRODH produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HSPOX2 PIG6 POX PRODH1 PRODH2 TP53I6; p53-induced gene 6 protein; proline dehydrogenase (oxidase) 1; Proline dehydrogenase 1, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; tumor protein p53 inducible protein 6
Gene Aliases: HSPOX2; PIG6; POX; POX2; PRODH; PRODH1; PRODH2; TP53I6
UniProt ID: (Human) O43272
Entrez Gene ID: (Human) 5625
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.