Invitrogen
Manufacturer Code:PA572540
Catalog # PIPA572540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 130-155 amino acids from the Central region of human PRODH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPOX2 PIG6 POX PRODH1 PRODH2 SCZD4 TP53I6 p53-induced gene 6 protein proline dehydrogenase 1 mitochondrial proline dehydrogenase mitochondrial proline oxidase 2 proline oxidase mitochondrial tumor protein p53 inducible protein 6; p53-induced gene 6 protein; proline dehydrogenase (oxidase) 1; Proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; tumor protein p53 inducible protein 6
Gene Aliases: HSPOX2; PIG6; POX; POX2; Pro-1; Pro1; PRODH; PRODH1; PRODH2; TP53I6; Ym24d07
UniProt ID: (Human) O43272, (Mouse) Q9WU79
Entrez Gene ID: (Human) 5625, (Mouse) 19125
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.