Invitrogen
Manufacturer Code:PA572297
Catalog # PIPA572297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 429-461 amino acids from human PRODH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPOX2 PIG6 POX PRODH1 PRODH2 SCZD4 TP53I6 p53-induced gene 6 protein proline dehydrogenase 1 mitochondrial proline dehydrogenase mitochondrial proline oxidase 2 proline oxidase mitochondrial tumor protein p53 inducible protein 6; Proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; Proline oxidase
Gene Aliases: Pro-1; Pro1; Prodh; Ym24d07
UniProt ID: (Mouse) Q9WU79
Entrez Gene ID: (Mouse) 19125
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.