Novus Biologicals
Manufacturer Code:NB1002832
Catalog # NB1002832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ERDGSGTNKRDKQYQ corresponding to internal region according to NP_057419.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.5.99.8 FLJ33744 HSPOX2 mitochondrial p53-induced gene 6 protein PIG6MGC148078 POX2 PRODH2MGC148079 proline dehydrogenase (oxidase) 1 proline dehydrogenase (proline oxidase ) Proline oxidase proline oxidase mitochondrial SCZD4 tumor protein p53 inducible protein 6; p53-induced gene 6 protein; proline dehydrogenase (oxidase) 1; Proline dehydrogenase 1, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; tumor protein p53 inducible protein 6
Gene Aliases: HSPOX2; PIG6; POX; POX2; PRODH; PRODH1; PRODH2; TP53I6
UniProt ID: (Human) O43272
Entrez Gene ID: (Human) 5625
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.