Proteintech
Manufacturer Code:229801AP150UL
Catalog # 50-173-3974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRODH Fusion Protein Ag18429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPOX2 p53 induced gene 6 protein PIG6 POX POX2 PRODH PRODH1 PRODH2 Proline oxidase Proline oxidase 2 SCZD4 TP53I6; p53-induced gene 6 protein; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); Proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; tumor protein p53 inducible protein 6
Gene Aliases: HSPOX2; PIG6; POX; POX2; Pro-1; Pro1; PRODH; PRODH1; PRODH2; TP53I6; Ym24d07
UniProt ID: (Human) O43272, (Mouse) Q9WU79
Entrez Gene ID: (Human) 5625, (Mouse) 19125, (Rat) 680409
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.