Proteintech
Manufacturer Code:125571AP150UL
Catalog # 50-172-8064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRMT8 Fusion Protein Ag3166 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginine methyltransferase 8; heterogeneous nuclear ribonucleoprotein methyltransferase-like 4; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4; HMT1 hnRNP methyltransferase-like 3; HRMT1L3 HRMT1L4 PRMT8; protein arginine N-methyltransferase 4; Protein arginine N-methyltransferase 8
Gene Aliases: HRMT1L3; HRMT1L4; PRMT8
UniProt ID: (Human) Q9NR22, (Mouse) Q6PAK3
Entrez Gene ID: (Human) 56341, (Mouse) 381813, (Rat) 688502
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.