Invitrogen
Manufacturer Code:720143
Catalog # PI720143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PRMT7 (aa 301-566) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Myelin basic protein]-arginine N-methyltransferase PRMT7; Histone-arginine N-methyltransferase PRMT7; myelin basic protein-arginine N-methyltransferase; Protein arginine N-methyltransferase 7; Protein arginine N-methyltransferase 7 KIAA1933 FLJ10640
Gene Aliases: KIAA1933; PRMT7
UniProt ID: (Human) Q9NVM4
Entrez Gene ID: (Human) 54496
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.