Invitrogen
Manufacturer Code:MA125470
Catalog # PIMA125470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PRMT5-21 |
Immunogen | Fusion of mouse myeloma cells and splenocytes from mice immunized with PRMT5 containing the C-terminal region of the Human PRMT5 (residues 315-637). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 72 kDa ICln-binding protein; Histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5 IBP72 JBP1 SKB1 SKB1Hs; Jak-binding protein 1; PRMT5; protein arginine methyltransferase 5; Protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; Shk1 kinase-binding protein 1 homolog; SKB1 homolog
Gene Aliases: HRMT1L5; I79_013398; IBP72; JBP1; PRMT5; SKB1; SKB1Hs
UniProt ID: (Human) O14744, (Bovine) A7YW45, (Mouse) Q8CIG8
Entrez Gene ID: (Human) 10419, (Bovine) 515594, (Dog) 480242, (Mouse) 27374, (Rat) 364382, (Chinese hamster) 100768836
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.