Novus Biologicals
Manufacturer Code:NBP219937
Catalog # NBP219937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PRMT5. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 72 kDa ICln-binding protein; EC 2.1.1.- EC 2.1.1.125 Histone-arginine N-methyltransferase PRMT5 HMT1 hnRNP methyltransferase-like 572 kDa ICln-binding protein HRMT1L5 IBP72 Jak-binding protein 1 JBP1 protein arginine methyltransferase 5 protein arginine N-methyltransferase 5 Shk1 kinase-binding protein 1 homolog SKB1 skb1 (S. pombe) homolog SKB1 homolog (S. pombe) SKB1HsSKB1 homolog; Histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; Jak-binding protein 1; PRMT5; Protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; Shk1 kinase-binding protein 1 homolog; SKB1 homolog
Gene Aliases: HRMT1L5; IBP72; JBP1; PRMT5; SKB1; SKB1Hs
UniProt ID: (Human) O14744
Entrez Gene ID: (Human) 10419
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.