Invitrogen
Manufacturer Code:MA125468
Catalog # PIMA125468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunodiffusion (IDF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PRMT1-171 |
Immunogen | Full length PRMT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANM1 HCP1 HRMT1L2 IR1B4; arginine N-methyltransferase 1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; Histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; Interferon receptor 1-bound protein 4; protein arginine methyltransferase 1; Protein arginine N-methyltransferase 1
Gene Aliases: 6720434D09Rik; ANM1; AW214366; HCP1; HMT2; HRMT1L2; IR1B4; Mrmt1; PRMT1
UniProt ID: (Human) Q99873, (Mouse) Q9JIF0, (Rat) Q63009
Entrez Gene ID: (Dog) 476411, (Bovine) 520388, (Human) 3276, (Mouse) 15469, (Chinese hamster) 100765197, (Rat) 60421
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.