Invitrogen
Manufacturer Code:MA535523
Catalog # PIMA535523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1066 |
Immunogen | A synthesized peptide derived from human PRMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 6720434D09Rik ANM1 AW214366 BOS_17676 HCP1 Histone-arginine N-methyltransferase PRMT1 HMT2 Hrmt1l2 Interferon receptor 1-bound protein 4 IR1B4 Mrmt1 PRMT1 Protein arginine N-methyltransferase 1; arginine N-methyltransferase 1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; Histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; Interferon receptor 1-bound protein 4; Protein arginine N-methyltransferase 1
Gene Aliases: 6720434D09Rik; ANM1; AW214366; HCP1; HMT2; HRMT1L2; IR1B4; Mrmt1; PRMT1
UniProt ID: (Human) Q99873, (Mouse) Q9JIF0
Entrez Gene ID: (Human) 3276, (Mouse) 15469
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.