Novus Biologicals
Manufacturer Code:NBP243744
Catalog # NBP243744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 779 |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PRMT1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANM1 EC 2.1.1 EC 2.1.1.- EC 2.1.1.125 EC 2.1.1.77 HCP1 Histone-arginine N-methyltransferase PRMT1 HMT1 (hnRNP methyltransferase S. cerevisiae)-like 2 HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae) HMT2 HRMT1L2HMT1 hnRNP methyltransferase-like 2 Interferon receptor 1-bound protein 4 IR1B4protein arginine N-methyltransferase 1 protein arginine methyltransferase 1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; Histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; Interferon receptor 1-bound protein 4; Protein arginine N-methyltransferase 1
Gene Aliases: ANM1; HCP1; HMT2; HRMT1L2; IR1B4; PRMT1
UniProt ID: (Human) Q99873
Entrez Gene ID: (Human) 3276
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.