Invitrogen
Manufacturer Code:PA130275
Catalog # PIPA130275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide AKNNRDLDFT IDLDFKG corresponding to amino acids 331-347 of Human PRMT 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANM1 HCP1 HRMT1L2 IR1B4; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; Histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; Interferon receptor 1-bound protein 4; Protein arginine N-methyltransferase 1
Gene Aliases: ANM1; HCP1; HMT2; HRMT1L2; IR1B4; PRMT1
UniProt ID: (Human) Q99873
Entrez Gene ID: (Human) 3276
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.