Invitrogen
Manufacturer Code:PA563293
Catalog # PIPA563293
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PRM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic nuclear protein HPI1; Basic nuclear protein HPI2; Basic nuclear protein HPS1; Basic nuclear protein HPS2; cancer/testis antigen family 94, member 2; P2'; P2''; Protamine-2; Sperm histone HP2; Sperm histone HP3; Sperm histone HP4; Sperm histone P2; Sperm protamine P2; Sperm protamine P3; Sperm protamine P4; testicular secretory protein Li 40
Gene Aliases: CT94.2; PRM2
UniProt ID: (Human) P04554
Entrez Gene ID: (Human) 5620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.