Proteintech
Manufacturer Code:145001AP150UL
Catalog # 50-172-9494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRM2 Fusion Protein Ag5952 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic nuclear protein HPI1; Basic nuclear protein HPI2; Basic nuclear protein HPS1; Basic nuclear protein HPS2; cancer/testis antigen family 94, member 2; P2 P2 Sperm histone HP3 PRM2 protamine 2 Sperm histone P2 Sperm protamine P2 Sperm protamine P3; P2'; P2''; PP2-A; PP2-B; PP2-C; PP2-D; Protamine mP2; Protamine-2; Sperm histone HP2; Sperm histone HP3; Sperm histone HP4; Sperm histone P2; Sperm protamine P2; Sperm protamine P3; Sperm protamine P4; sperm protamine-P2; testicular secretory protein Li 40
Gene Aliases: AI528784; CT94.2; Prm-2; PRM2
UniProt ID: (Human) P04554, (Mouse) P07978
Entrez Gene ID: (Human) 5620, (Mouse) 19119
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.