Invitrogen
Manufacturer Code:PA5101865
Catalog # PIPA5101865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRKX corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP-dependent protein kinase catalytic subunit PRKX; cAMP-dependent protein kinase catalytic subunit PRKX Pkare PKA-related protein kinase PKX1 PrKX protein kinase PKX1 protein kinase X protein kinase X-linked protein kinase X-linked putative serine/threonine kinase serine/threonine-protein kinase PRKX; PKA-related protein kinase; PrKX; Protein kinase PKX1; protein kinase X; protein kinase X-linked; putative serine/threonine kinase; serine/threonine-protein kinase PRKX
Gene Aliases: Pkare; PKX1; PRKX
UniProt ID: (Human) P51817, (Mouse) Q922R0
Entrez Gene ID: (Human) 5613, (Mouse) 19108, (Rat) 501563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.