OriGene
Manufacturer Code:TA807769
Catalog # 50-168-1752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI15E6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 40-320 of human PRKG2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cGK 2; cGKII; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; PRKGR2 cGK2 cGKII; testicular tissue protein Li 99
Gene Aliases: cGK2; cGKII; PRKG2; PRKGR2
UniProt ID: (Human) Q13237
Entrez Gene ID: (Human) 5593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.