OriGene
Manufacturer Code:TA501208
Catalog # 50-167-2352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9A4 |
Immunogen | Full length human recombit protein of human PRKG1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AAT8 PKG PRKG1B PRKGR1B cGK cGK 1 cGK1 cGKI cGKI-BETA cGKI-alpha; cGK 1; cGKI; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; protein kinase, cGMP-dependent, regulatory, type I, beta
Gene Aliases: AAT8; cGK; cGK 1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; PKG; PRKG1; PRKG1B; PRKGR1A; PRKGR1B
UniProt ID: (Human) Q13976
Entrez Gene ID: (Human) 5592
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.