Proteintech
Manufacturer Code:216461AP150UL
Catalog # 50-173-3533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRKG1 Fusion Protein Ag16333 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGK 1; cGK 1 cGK1 CGKI cGKI alpha cGKI BETA PGK PKG PRKG1 PRKG1B PRKGR1A PRKGR1B Protein Kinase G1 alpha; cGK1; cGKI; cGMP dependent protein kinase type 1; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type I
Gene Aliases: AAT8; AW125416; cGK; cGK 1; cGK1; CGKI; cGKI-alpha; cGKI-BETA; Gm19690; PKG; Pkgi; PRKG1; PRKG1B; PRKGR1A; PRKGR1B
UniProt ID: (Human) Q13976, (Mouse) P0C605
Entrez Gene ID: (Human) 5592, (Mouse) 19091, (Rat) 54286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.