Abnova
Manufacturer Code:PAB26735
Catalog # 89-128-375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding T2609 of human PRKDC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PRKDC.
For Research Use Only
Protein Aliases: DNA-dependent protein kinase catalytic subunit; DNA-PK catalytic subunit; DNA-PKcs; DNAPK; DNPK1; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; p350; p460; XRCC7
Gene Aliases: DNA-PKcs; DNAPK; DNPK1; HYRC; HYRC1; IMD26; p350; PRKDC; XRCC7
UniProt ID: (Human) P78527
Entrez Gene ID: (Human) 5591
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.